missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-pyrrolidone 98.0+%, TCI America™

Catalog No. M040610G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M040610G 10 g
M040625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M040610G Supplier TCI America Supplier No. M040610G

Chemical Identifiers

CAS 108-27-0
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00005273
InChI Key YVIVRJLWYJGJTJ-UHFFFAOYSA-N
Synonym 5-Methyl-2-pyrrolidinone
PubChem CID 66055
IUPAC Name 5-methylpyrrolidin-2-one
SMILES CC1CCC(=O)N1

Specifications

Color White-Yellow
Boiling Point 142°C
Quantity 10 g
Formula Weight 99.13
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 5-Methyl-2-pyrrolidone