missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-2-phenyl-3-furyl)methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 771572-29-3
Molecular Formula C12H13NO
Molecular Weight (g/mol) 187.24
MDL Number MFCD06213531
InChI Key PQZVRVFUCJMCRZ-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-3-furyl methylamine, 5-methyl-2-phenylfuran-3-yl methanamine, 1-5-methyl-2-phenylfuran-3-yl methanamine, 3-furanmethanamine,5-methyl-2-phenyl, 5-methyl-2-phenyl-3-furyl methanamine, 5-methyl-2-phenylfur-3-yl methylamine
PubChem CID 24229505
IUPAC Name (5-methyl-2-phenylfuran-3-yl)methanamine
SMILES CC1=CC(CN)=C(O1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC49713CB
View Documents
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC49713DA
View Documents
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

771572-29-3
187.24
PQZVRVFUCJMCRZ-UHFFFAOYSA-N
24229505
CC1=CC(CN)=C(O1)C1=CC=CC=C1
C12H13NO
MFCD06213531
5-methyl-2-phenyl-3-furyl methylamine, 5-methyl-2-phenylfuran-3-yl methanamine, 1-5-methyl-2-phenylfuran-3-yl methanamine, 3-furanmethanamine,5-methyl-2-phenyl, 5-methyl-2-phenyl-3-furyl methanamine, 5-methyl-2-phenylfur-3-yl methylamine
(5-methyl-2-phenylfuran-3-yl)methanamine

Specifications

771572-29-3
100.0
C12H13NO
5-methyl-2-phenyl-3-furyl methylamine, 5-methyl-2-phenylfuran-3-yl methanamine, 1-5-methyl-2-phenylfuran-3-yl methanamine, 3-furanmethanamine,5-methyl-2-phenyl, 5-methyl-2-phenyl-3-furyl methanamine, 5-methyl-2-phenylfur-3-yl methylamine
CC1=CC(CN)=C(O1)C1=CC=CC=C1
187.24
187.24
(5-Methyl-2-phenyl-3-furyl)methylamine
96.5
97%
MFCD06213531
PQZVRVFUCJMCRZ-UHFFFAOYSA-N
(5-methyl-2-phenylfuran-3-yl)methanamine
24229505
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents