missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-2-phenyl-3-furyl)methanol, 95%, Thermo Scientific™

Catalog No. CC49709CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC49709CB 250mg Amber Glass Bottle
CC49709DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC49709CB Supplier Thermo Scientific Chemicals Supplier No. CC49709CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 183210-33-5
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD08435905
InChI Key VZFFMWAZPODJRX-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-3-furyl methanol, 5-methyl-2-phenylfuran-3-yl methanol, 3-furanmethanol,5-methyl-2-phenyl, 5-methyl-2-phenyl-3-furyl methan-1-ol
PubChem CID 15269738
IUPAC Name (5-methyl-2-phenylfuran-3-yl)methanol
SMILES CC1=CC(=C(O1)C2=CC=CC=C2)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 188.22
Percent Purity 95%
Chemical Name or Material (5-Methyl-2-phenyl-3-furyl)methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.