missing translation for 'onlineSavingsMsg'
Learn More

5-methyl-2-phenyl-2H-1,2,3-triazole-4-carbonyl chloride, Thermo Scientific™

$55.89 - $55.89

Chemical Identifiers

CAS 36401-55-5
Molecular Formula C10H8ClN3O
Molecular Weight (g/mol) 221.64
MDL Number MFCD00052545
InChI Key UJYBUZMRRLFXGM-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonyl chloride, 5-methyl-2-phenyl-1,2,3-triazole-4-carbonyl chloride, 2h-1,2,3-triazole-4-carbonylchloride, 5-methyl-2-phenyl, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbonyl chloride, 5-methyl-2-phenyl-4-triazolecarbonyl chloride, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbo nyl chloride, 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonylchloride
PubChem CID 2776447
IUPAC Name 5-methyl-2-phenyltriazole-4-carbonyl chloride
SMILES CC1=NN(N=C1C(Cl)=O)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC16202CB
SDS View Documents
Thermo Scientific Chemicals
CC16202CB
250 mg Amber Glass Bottle
Each for $55.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

36401-55-5
221.64
UJYBUZMRRLFXGM-UHFFFAOYSA-N
2776447
CC1=NN(N=C1C(Cl)=O)C1=CC=CC=C1
C10H8ClN3O
MFCD00052545
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonyl chloride, 5-methyl-2-phenyl-1,2,3-triazole-4-carbonyl chloride, 2h-1,2,3-triazole-4-carbonylchloride, 5-methyl-2-phenyl, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbonyl chloride, 5-methyl-2-phenyl-4-triazolecarbonyl chloride, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbo nyl chloride, 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonylchloride
5-methyl-2-phenyltriazole-4-carbonyl chloride

Specifications

36401-55-5
Amber Glass Bottle
MFCD00052545
5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonyl chloride, 5-methyl-2-phenyl-1,2,3-triazole-4-carbonyl chloride, 2h-1,2,3-triazole-4-carbonylchloride, 5-methyl-2-phenyl, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbonyl chloride, 5-methyl-2-phenyl-4-triazolecarbonyl chloride, 5-methyl-2-phenyl-2h-1,2,3 triazole-4-carbo nyl chloride, 5-methyl-2-phenyl-2h-1,2,3-triazole-4-carbonylchloride
CC1=NN(N=C1C(Cl)=O)C1=CC=CC=C1
221.64
221.65
5-Methyl-2-phenyl-2H-1, 2, 3-triazole-4-carbonyl chloride
97%
C10H8ClN3O
250 mg
UJYBUZMRRLFXGM-UHFFFAOYSA-N
5-methyl-2-phenyltriazole-4-carbonyl chloride
2776447
97%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'