missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-2-phenyl-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 250mg |
| Formula Weight | 203.2 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Methyl-2-phenyl-1, 3-oxazole-4-carboxylic acid |