missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-2-hexanol 98.0+%, TCI America™
Supplier: TCI America M166625G
Specifications
| 5-Methyl-2-hexanol | |
| Colorless | |
| C7H16O | |
| 25 g | |
| 5-methyl-2-hexanol, 2-hexanol, 5-methyl, 2-methyl-5-hexanol, 2-hexanol,5-methyl, 5-methylhexanol-2, acmc-1b03m, zdvjgwxfxgjsiu-uhfffaoysa | |
| CC(C)CCC(C)O | |
| 116.20 | |
| 116.20 | |
| Liquid |
| 627-59-8 | |
| 150°C | |
| MFCD00004558 | |
| 1987 | |
| ZDVJGWXFXGJSIU-UHFFFAOYNA-N | |
| 5-methylhexan-2-ol | |
| 12323 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 627-59-8 | |
| 116.20 | |
| ZDVJGWXFXGJSIU-UHFFFAOYNA-N | |
| 12323 | |
| CC(C)CCC(C)O |
| C7H16O | |
| MFCD00004558 | |
| 5-methyl-2-hexanol, 2-hexanol, 5-methyl, 2-methyl-5-hexanol, 2-hexanol,5-methyl, 5-methylhexanol-2, acmc-1b03m, zdvjgwxfxgjsiu-uhfffaoysa | |
| 5-methylhexan-2-ol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No