missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-hepten-4-one, predominantly trans, 99%

Catalog No. p-7026369
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2233504 2 g
AAB2233509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2233504 Supplier Thermo Scientific Chemicals Supplier No. B2233504
Only null left

CAS: 81925-81-7 | C8H14O | 126.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81925-81-7
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00792509
InChI Key ARJWAURHQDJJAC-GQCTYLIASA-N
Synonym 5-methyl-2-hepten-4-one, filbertone, 5-methylhept-2-en-4-one, +/--filbertone, hazelnut ketone, e-5-methyl-2-hepten-4-one, 5-methyl-e-2-hepten-4-one, fema no. 3761, e-5-methylhept-2-en-4-one, 2e-5-methyl-2-hepten-4-one
PubChem CID 5362588
IUPAC Name (E)-5-methylhept-2-en-4-one
SMILES CCC(C)C(=O)C=CC

Specifications

Boiling Point 169°C to 171°C
Flash Point 55°C (131°F)
Refractive Index 1.444
Quantity 2 g
UN Number UN1224
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 5-Methyl-2-hepten-4-one, predominantly trans
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only