missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-furanmethanamine, 98%

Catalog No. AC268940050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC268940050 5 g Glass bottle
AC268940250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC268940050 Supplier Thermo Scientific Chemicals Supplier No. 268940050

CAS: 14003-16-8 | C6H9NO | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14003-16-8
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD00143471
InChI Key YSEAGSCGERFGBL-UHFFFAOYSA-N
Synonym 5-methylfurfurylamine, 5-methylfuran-2-yl methanamine, 5-methyl-2-furanmethanamine, 2-aminomethyl-5-methylfuran, 5-methyl-2-furyl methylamine, 2-furanmethanamine, 5-methyl, 5-methyl-2-furyl methyl amine, 1-5-methylfuran-2-yl methanamine, pubchem6995, acmc-209cjz
PubChem CID 2724683
IUPAC Name (5-methylfuran-2-yl)methanamine
SMILES CC1=CC=C(CN)O1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -22°C
Color Yellow
Density 1.0100g/mL
Boiling Point 57°C (13.0 Mbar)
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4860 to 1.488
Quantity 5 g
Specific Gravity 1.01
Solubility Information Solubility in water: >1000g/L (20°C)
Formula Weight 111.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-furanmethanamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only