missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-furaldehyde 98.0+%, TCI America™

Catalog No. p-7132080
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
M025425ML 25 mL
M0254500ML 500 mL
2 options

Catalog No. M025425ML

Supplier: TCI America M025425ML

Only null left

Chemical Identifiers

CAS 620-02-0
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00003232
InChI Key OUDFNZMQXZILJD-UHFFFAOYSA-N
Synonym 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
PubChem CID 12097
ChEBI CHEBI:2091
IUPAC Name 5-methylfuran-2-carbaldehyde
SMILES CC1=CC=C(O1)C=O

Specifications

Color Yellow
Boiling Point 187°C
Quantity 25 mL
Formula Weight 110.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-furaldehyde
EINECSNumber (5)-0041
RTECSNumber LT7032500
TSCA Yes