missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-2-furaldehyde 98.0+%, TCI America™

Catalog No. M025425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M025425ML 25 mL
M0254500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M025425ML Supplier TCI America Supplier No. M025425ML

Chemical Identifiers

CAS 620-02-0
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00003232
InChI Key OUDFNZMQXZILJD-UHFFFAOYSA-N
Synonym 5-methylfurfural, 5-methyl-2-furaldehyde, 5-methyl furfural, 5-methyl-2-furfural, 2-furancarboxaldehyde, 5-methyl, 5-methylfuran-2-al, 5-methyl-2-furancarboxaldehyde, 5-methylfurfuraldehyde, 2-furaldehyde, 5-methyl, 2-formyl-5-methylfuran
PubChem CID 12097
ChEBI CHEBI:2091
IUPAC Name 5-methylfuran-2-carbaldehyde
SMILES CC1=CC=C(O1)C=O

Specifications

Color Yellow
Boiling Point 187°C
Quantity 25 mL
Formula Weight 110.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-furaldehyde
EINECSNumber (5)-0041
RTECSNumber LT7032500
TSCA Yes