missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-2'-deoxycytidine, MP Biomedicals™
5-Methyl-2'-deoxycytidine, White Powder
$108.63 - $108.63
Chemical Identifiers
| CAS | 838-07-3 |
|---|---|
| MDL Number | MFCD00006549 |
| InChI Key | LUCHPKXVUGJYGU-XLPZGREQSA-N |
| PubChem CID | 440055 |
| ChEBI | CHEBI:47876 |
| IUPAC Name | 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one |
| SMILES | CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19888310
|
MP Biomedicals Inc
0219888310 |
10 mg |
N/A
|
|
|||||
Description
- 5-Methyl-2'-deoxycytidine in single-stranded DNA can act in cis to signal de novo DNA methylation.
- It has been used in epigenetics research to investigate the dynamics of DNA methylation pattern in the control of gene expression.
- A cytidine analog and an isostere of thymidine.
- 5-Methyl-2'-deoxycytidine is a pyrimidine nucleoside that when incorporated into single-stranded DNA can act in cis to signal de novo DNA methylation.
Chemical Identifiers
| 838-07-3 | |
| LUCHPKXVUGJYGU-XLPZGREQSA-N | |
| CHEBI:47876 | |
| CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O |
| MFCD00006549 | |
| 440055 | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one |
Specifications
| 838-07-3 | |
| White | |
| C10H15N5O3 | |
| MFCD00006549 | |
| 5-methyl-2'-deoxycytidine, 5-methyldeoxycytidine, 2'-deoxy-5-methylcytidine, unii-b200gv71qm, cytidine, 2'-deoxy-5-methyl, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-methylpyrimidin-2 1h-one, 5-mdc, 1-2-deoxy-beta-d-ribofuranosyl-5-methylcytosine, 5-methyl-2-deoxycytidine, 4-25-00-03727 beilstein handbook reference | |
| LUCHPKXVUGJYGU-XLPZGREQSA-N | |
| CC1=CN(C(=O)N=C1N)C2CC(C(O2)CO)O | |
| 241.2 | |
| CHEBI:47876 | |
| 8.85E-12mmHg at 25°C (lit.) | |
| 5-Methyl-2'-deoxycytidine |
| 217°C to 219°C (Literature) | |
| >99.5% | |
| 1.694 (lit.) | |
| 10 mg | |
| Soluble in 1 N NaOH. | |
| [α]25/D +63°, c = 1 in 1 M NaOH (lit.) | |
| 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidin-2-one | |
| 440055 | |
| 241.2 | |
| Powder | |
| 99.6 area% (HPLC) |
Safety and Handling
RTECSNumber : HA3860000
storageNote1 : Store at Room Temperature (15-30°C).