missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1H-tetrazole, 97%

Catalog No. AAH6024806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6024806 5 g
AAH6024814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6024806 Supplier Thermo Scientific Chemicals Supplier No. H6024806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4076-36-2 | C2H4N4 | 84.082 g/mol

5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1H-tetrazole is used in the preparation of novel antifungal agents based derived from N-iodopropargylazoles and N-triiodoallylazoles. 5-Methyl Tetrazole also in the preparation of novel quinoline derivatives against mycobacterium tuberculosis.

Solubility
Soluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.

Chemical Identifiers

CAS 4076-36-2
Molecular Formula C2H4N4
Molecular Weight (g/mol) 84.082
MDL Number MFCD00129971
InChI Key XZGLNCKSNVGDNX-UHFFFAOYSA-N
Synonym 5-methyltetrazole, 5-methyl-1h-tetrazole, 5-methyl tetrazole, 1h-tetrazole, 5-methyl, 5-methyl-1h-tertazole, tetrazole, 5-methyl, 5-methyl-1h-1,2,3,4-tetrazole, 5-methyl-2h-1,2,3,4-tetrazole, 2h-tetrazole, 5-methyl, 5-methyl-1h-1,2,3,4-tetraazole
PubChem CID 138107
IUPAC Name 5-methyl-2H-tetrazole
SMILES CC1=NNN=N1

Specifications

Melting Point 142°C to 146°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 84.08
Percent Purity 97%
Chemical Name or Material 5-Methyl-1H-tetrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-746-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only