missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-1H-benzotriazole 99.0+%, TCI America™
$117.43 - $881.70
Chemical Identifiers
| CAS | 136-85-6 |
|---|---|
| Molecular Formula | C7H7N3 |
| Molecular Weight (g/mol) | 133.15 |
| MDL Number | MFCD00005702 |
| InChI Key | LRUDIIUSNGCQKF-UHFFFAOYSA-N |
| Synonym | 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1 |
| PubChem CID | 8705 |
| ChEBI | CHEBI:83455 |
| IUPAC Name | 5-methyl-2H-1,2,3-benzotriazole |
| SMILES | CC1=CC2=NNN=C2C=C1 |
Chemical Identifiers
| 136-85-6 | |
| 133.15 | |
| LRUDIIUSNGCQKF-UHFFFAOYSA-N | |
| 8705 | |
| 5-methyl-2H-1,2,3-benzotriazole |
| C7H7N3 | |
| MFCD00005702 | |
| 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1 | |
| CHEBI:83455 | |
| CC1=CC2=NNN=C2C=C1 |
Specifications
| 136-85-6 | |
| 323°C | |
| MFCD00005702 | |
| 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1 | |
| CC1=CC2=NNN=C2C=C1 | |
| 133.15 | |
| CHEBI:83455 | |
| ≥99.0% (T) | |
| 5-Methyl-1H-benzotriazole |
| 83°C | |
| C7H7N3 | |
| 25 g | |
| LRUDIIUSNGCQKF-UHFFFAOYSA-N | |
| 5-methyl-2H-1,2,3-benzotriazole | |
| 8705 | |
| 133.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-0539
RTECSNumber : DM1400000
TSCA : Yes
Recommended Storage : Refrigerator