missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1H-benzotriazole 99.0+%, TCI America™

Catalog No. M0249500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0249500G 500 g
M024925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0249500G Supplier TCI America Supplier No. M0249500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 136-85-6
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005702
InChI Key LRUDIIUSNGCQKF-UHFFFAOYSA-N
Synonym 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1
PubChem CID 8705
ChEBI CHEBI:83455
IUPAC Name 5-methyl-2H-1,2,3-benzotriazole
SMILES CC1=CC2=NNN=C2C=C1

Specifications

Melting Point 83°C
Boiling Point 323°C
Quantity 500 g
Formula Weight 133.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-1H-benzotriazole
EINECSNumber (5)-0539
RTECSNumber DM1400000
TSCA Yes
Recommended Storage Refrigerator