Learn More
5-Methyl-1H-benzotriazole, 98+%
CAS: 136-85-6 | C7H7N3 | 133.15 g/mol
$128.86 - $1675.27
Chemical Identifiers
| CAS | 136-85-6 |
|---|---|
| Molecular Formula | C7H7N3 |
| Molecular Weight (g/mol) | 133.15 |
| MDL Number | MFCD00005702 |
| InChI Key | LRUDIIUSNGCQKF-UHFFFAOYSA-N |
| Synonym | 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1 |
| PubChem CID | 8705 |
| ChEBI | CHEBI:83455 |
| IUPAC Name | 5-methyl-2H-benzotriazole |
| SMILES | CC1=CC2=NNN=C2C=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1061514
|
Thermo Scientific Chemicals
A1061514 |
25 g |
Each for $128.86
|
|
|||||
|
AAA1061522
|
Thermo Scientific Chemicals
A1061522 |
100 g |
Each for $372.48
|
|
|||||
|
AAA1061536
|
Thermo Scientific Chemicals
A1061536 |
500 g |
Each for $1,675.27
|
|
|||||
Description
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.
Solubility
Soluble in chloroform, methanol, water (6.0 g/L at 25°C), ethanol, and toluene (slightly).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 136-85-6 | |
| 133.15 | |
| LRUDIIUSNGCQKF-UHFFFAOYSA-N | |
| 8705 | |
| 5-methyl-2H-benzotriazole |
| C7H7N3 | |
| MFCD00005702 | |
| 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1 | |
| CHEBI:83455 | |
| CC1=CC2=NNN=C2C=C1 |
Specifications
| 136-85-6 | |
| 5.3 | |
| Odorless | |
| MFCD00005702 | |
| 116658 | |
| Soluble in chloroform,methanol,water (6.0g/L at 25°C),ethanol,and toluene (slightly). | |
| CC1=CC2=NNN=C2C=C1 | |
| 133.15 | |
| CHEBI:83455 | |
| ≥98% |
| 77°C to 84°C | |
| 210°C to 212°C (12 mmHg) | |
| C7H7N3 | |
| 25 g | |
| 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1 | |
| LRUDIIUSNGCQKF-UHFFFAOYSA-N | |
| 5-methyl-2H-benzotriazole | |
| 8705 | |
| 133.15 | |
| 5-Methyl-1H-benzotriazole |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 205-265-8
RTECSNumber : DM1400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only