missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride, 95+%, Thermo Scientific™
Chemical Identifiers
| CAS | 342405-38-3 |
|---|---|
| Molecular Formula | C10H9ClN2O2S |
| Molecular Weight (g/mol) | 256.70 |
| MDL Number | MFCD02681919 |
| InChI Key | IRJCRNZTDRODFQ-UHFFFAOYSA-N |
| Synonym | 5-methyl-1-phenyl-1h-pyrazole-4-sulfonyl chloride, 5-methyl-1-phenyl-1h-pyrazole-4-sulphonyl chloride, 5-methyl-1-phenyl-pyrazole-4-sulfonyl chloride, 4-chlorosulphonyl-1-phenyl-1h-pyrazole, 5-methyl-1-phenyl-4-pyrazolesulfonyl chloride, 5-methyl-1-phenylpyrazol-4-sulfonyl chloride, chloro 5-methyl-1-phenylpyrazol-4-yl sulfone, 1h-pyrazole-4-sulfonylchloride, 5-methyl-1-phenyl, 4-chlorosulphonyl-5-methyl-1h-pyrazol-1-yl benzene |
| PubChem CID | 2776435 |
| IUPAC Name | 5-methyl-1-phenylpyrazole-4-sulfonyl chloride |
| SMILES | CC1=C(C=NN1C1=CC=CC=C1)S(Cl)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
CC15503DA
|
N/A | 1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC15503DE
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 342405-38-3 | |
| 256.70 | |
| IRJCRNZTDRODFQ-UHFFFAOYSA-N | |
| 2776435 | |
| CC1=C(C=NN1C1=CC=CC=C1)S(Cl)(=O)=O |
| C10H9ClN2O2S | |
| MFCD02681919 | |
| 5-methyl-1-phenyl-1h-pyrazole-4-sulfonyl chloride, 5-methyl-1-phenyl-1h-pyrazole-4-sulphonyl chloride, 5-methyl-1-phenyl-pyrazole-4-sulfonyl chloride, 4-chlorosulphonyl-1-phenyl-1h-pyrazole, 5-methyl-1-phenyl-4-pyrazolesulfonyl chloride, 5-methyl-1-phenylpyrazol-4-sulfonyl chloride, chloro 5-methyl-1-phenylpyrazol-4-yl sulfone, 1h-pyrazole-4-sulfonylchloride, 5-methyl-1-phenyl, 4-chlorosulphonyl-5-methyl-1h-pyrazol-1-yl benzene | |
| 5-methyl-1-phenylpyrazole-4-sulfonyl chloride |
Specifications
| 342405-38-3 | |
| C10H9ClN2O2S | |
| 5-methyl-1-phenyl-1h-pyrazole-4-sulfonyl chloride, 5-methyl-1-phenyl-1h-pyrazole-4-sulphonyl chloride, 5-methyl-1-phenyl-pyrazole-4-sulfonyl chloride, 4-chlorosulphonyl-1-phenyl-1h-pyrazole, 5-methyl-1-phenyl-4-pyrazolesulfonyl chloride, 5-methyl-1-phenylpyrazol-4-sulfonyl chloride, chloro 5-methyl-1-phenylpyrazol-4-yl sulfone, 1h-pyrazole-4-sulfonylchloride, 5-methyl-1-phenyl, 4-chlorosulphonyl-5-methyl-1h-pyrazol-1-yl benzene | |
| CC1=C(C=NN1C1=CC=CC=C1)S(Cl)(=O)=O | |
| 256.70 | |
| 256.71 | |
| 5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride |
| 95% | |
| MFCD02681919 | |
| IRJCRNZTDRODFQ-UHFFFAOYSA-N | |
| 5-methyl-1-phenylpyrazole-4-sulfonyl chloride | |
| 2776435 | |
| 95% |