missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC15504EA

 View more versions of this product

Catalog No. CC15504EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

5-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
97%
C11H10N2O
5-methyl-1-phenyl-1h-pyrazole-4-carbaldehyde, 5-methyl-1-phenyl-1h-pyrazole-4-carboxaldehyde, acmc-20am2d, 1h-pyrazole-4-carboxaldehyde, 5-methyl-1-phenyl, 1h-pyrazole-4-carboxaldehyde,5-methyl-1-phenyl
CC1=C(C=NN1C2=CC=CC=C2)C=O
186.214
186.21
98700-50-6
Amber Glass Bottle
10g
IIEFFVFWBUGUTI-UHFFFAOYSA-N
5-methyl-1-phenylpyrazole-4-carbaldehyde
2776437
97%

Chemical Identifiers

98700-50-6
186.214
5-methyl-1-phenyl-1h-pyrazole-4-carbaldehyde, 5-methyl-1-phenyl-1h-pyrazole-4-carboxaldehyde, acmc-20am2d, 1h-pyrazole-4-carboxaldehyde, 5-methyl-1-phenyl, 1h-pyrazole-4-carboxaldehyde,5-methyl-1-phenyl
5-methyl-1-phenylpyrazole-4-carbaldehyde
C11H10N2O
IIEFFVFWBUGUTI-UHFFFAOYSA-N
2776437
CC1=C(C=NN1C2=CC=CC=C2)C=O
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions