missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-indanone, 97%

Catalog No. AAH6681406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6681406 5 g
AAH6681414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6681406 Supplier Thermo Scientific Chemicals Supplier No. H6681406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4593-38-8 | C10H10O | 146.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4593-38-8
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
MDL Number MFCD02179276
InChI Key KBHCTNGQJOEDDC-UHFFFAOYSA-N
Synonym 5-methyl-1-indanone, 5-methyl-2,3-dihydro-1h-inden-1-one, 5-methylindan-1-one, 5-methyl-indanone, 1h-inden-1-one, 2,3-dihydro-5-methyl, 5-methy1-2,3-dihydroinden-1-one, 5-methylindanone, pubchem8850, 1h-inden-1-one,2,3-dihydro-5-methyl
PubChem CID 10130031
IUPAC Name 5-methyl-2,3-dihydroinden-1-one
SMILES CC1=CC2=C(C=C1)C(=O)CC2

Specifications

Melting Point 69°C to 73°C
Quantity 5 g
Beilstein 119335
Formula Weight 146.19
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Methyl-1-indanone
Hazard Category H302-H319
Hazard Statement GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only