Learn More
5-Methyl-1-hexyne, 98%
CAS: 2203-80-7 | C7H12 | 96.173 g/mol
Supplier: Thermo Scientific Chemicals L0601414
Description
5-Methyl-1-hexyne is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Methyl-1-hexyne is used as pharmaceutical intermediates.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications
| 5-Methyl-1-hexyne | |
| 0.732 | |
| −4°C (24°F) | |
| 1.4075 | |
| 25 g | |
| 1731845 | |
| Not miscible in water. | |
| CC(C)CCC#C | |
| 96.173 | |
| 96.17 |
| 2203-80-7 | |
| 91°C to 93°C | |
| C7H12 | |
| MFCD00041610 | |
| UN3295 | |
| 5-methyl-1-hexyne, 1-hexyne, 5-methyl, isoheptyne, unii-lro6ea6hk1, lro6ea6hk1, isoamylacetylene, isohept-1-yne, acmc-1cg6t, hknanemucjgpms-uhfffaoysa | |
| HKNANEMUCJGPMS-UHFFFAOYSA-N | |
| 5-methylhex-1-yne | |
| 137490 | |
| 98% |
Chemical Identifiers
| 2203-80-7 | |
| 96.173 | |
| HKNANEMUCJGPMS-UHFFFAOYSA-N | |
| 137490 | |
| CC(C)CCC#C |
| C7H12 | |
| MFCD00041610 | |
| 5-methyl-1-hexyne, 1-hexyne, 5-methyl, isoheptyne, unii-lro6ea6hk1, lro6ea6hk1, isoamylacetylene, isohept-1-yne, acmc-1cg6t, hknanemucjgpms-uhfffaoysa | |
| 5-methylhex-1-yne |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H225-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only