missing translation for 'onlineSavingsMsg'
Learn More

5-methyl-1-[3-(trifluoromethyl)phenyl]-1H-1,2,3-triazole-4-carboxylic acid, Thermo Scientific™

Catalog No. CD04550CB
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CD04550CB 250mg
CD04550DA 1g
CD04550DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CD04550CB Supplier Thermo Scientific Chemicals Supplier No. CD04550CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 499771-21-0
Molecular Formula C11H8F3N3O2
Molecular Weight (g/mol) 271.199
InChI Key IRJLZHQBWAUKGA-UHFFFAOYSA-N
Synonym 5-methyl-1-3-trifluoromethyl phenyl-1h-1,2,3-triazole-4-carboxylic acid, 5-methyl-1-3-trifluoromethyl phenyl-1,2,3-triazole-4-carboxylic acid, maybridge1_002663, 5-methyl-1-3-trifluoromethyl phenyl-4-triazolecarboxylic acid, 5-methyl-1-3-trifluoromethyl phenyl triazole-4-carboxylic acid, 5-methyl-1-3-trifluoromethyl phenyl-1h-1,2,3-triazole-4carboxylic acid
PubChem CID 2804089
IUPAC Name 5-methyl-1-[3-(trifluoromethyl)phenyl]triazole-4-carboxylic acid
SMILES CC1=C(N=NN1C2=CC=CC(=C2)C(F)(F)F)C(=O)O

Specifications

Assay Percent Range 97%
Quantity 250mg
Formula Weight 271.2
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-[3-(trifluoromethyl)phenyl]-1H-1, 2, 3-triazole-4-carboxylic acid
Hazard Category GHS Signal Word: Warning