missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1,3-cyclohexanedione 98.0+%, TCI America™

Catalog No. M09455G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09455G 5 g
M09451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09455G Supplier TCI America Supplier No. M09455G

Chemical Identifiers

CAS 4341-24-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00010379
InChI Key DMIIMPQQPXUKOO-UHFFFAOYSA-N
PubChem CID 458095
IUPAC Name 5-methylcyclohexane-1,3-dione
SMILES CC1CC(=O)CC(=O)C1

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 5 g
Synonym 5-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 5-methyl, 5-methyl-cyclohexane-1,3-dione, pubchem14279, acmc-1aftb, 5-methylcyclohexane dione, 1,3-dioxo-5-methylcyclohexane, 1,3-cyclohexanedione,5-methyl, 5-methyl cyclohexane-1,3-dione
Formula Weight 126.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-1,3-cyclohexanedione