missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxytryptamine, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022850
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022850 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022850 Supplier Merck Emd Millipore Supplier No. BP1078

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 608-07-1
Melting Point 121°C to 123°C (literature)
Molecular Formula C11H14N2O
Linear Formula C11H14N2O
Quantity 25 mg
MDL Number MFCD00005662
Synonym 2-(5-Methoxyindol-3-yl)ethylamine; 3-(2-Aminoethyl)-5-methoxyindole; 5-MOT; 5-Methoxyindole-3-ethanamine; Deacetylmelatonin; NSC 56422
Formula Weight 190.24
Grade British Pharmacopoeia (BP) Reference Standard