missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxypsoralen, 98%

Catalog No. p-7077850
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460660010 1 g
AC460660050 5 g
AC460660250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC460660010 Supplier Thermo Scientific Chemicals Supplier No. 460660010
Only null left

CAS: 484-20-8 | C12H8O4 | 216.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 484-20-8
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
InChI Key BGEBZHIAGXMEMV-UHFFFAOYSA-N
Synonym bergapten, 5-methoxypsoralen, bergaptene, heraclin, majudin, 4-methoxy-7h-furo 3,2-g chromen-7-one, bergaptan, psoraderm, 5-mop, o-methylbergaptol
PubChem CID 2355
ChEBI CHEBI:18293
IUPAC Name 4-methoxyfuro[3,2-g]chromen-7-one
SMILES COC1=C2C=CC(=O)OC2=CC3=C1C=CO3

Specifications

Melting Point 190.0°C to 193.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 216.19
Percent Purity 98%
Chemical Name or Material 5-Methoxypsoralen
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
In case of inadequate ventilation wear respiratory protection.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for brea
EINECSNumber 207-604-5

RUO – Research Use Only