missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methoxypsoralen, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 190.0°C to 193.0°C |
| Loss on Drying | 2% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Quantity | 25 g |
| Formula Weight | 216.19 |
| Percent Purity | 98% |
| Chemical Name or Material | 5-Methoxypsoralen |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause genetic defects. May cause cancer. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. In case of inadequate ventilation wear respiratory protection. IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for brea |
| EINECSNumber | 207-604-5 |
RUO – Research Use Only