missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxypiperonal, 97%

CAS: 5780-07-4 | C9H8O4 | 180.16 g/mol

Supplier:  Thermo Scientific Chemicals L0821306

 View more versions of this product

Catalog No. AAL0821306

Only null left
Explore available promotions

Description

Description

5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with air, oxidizing agents. Store under dry inert gas. Store away from air.
Specifications

Specifications

5-Methoxypiperonal
126°C to 132°C
MFCD00016900
384077
myristicin aldehyde, 3-methoxy-4,5-methylenedioxybenzaldehyde, 3-methoxypiperonal, 1,3-benzodioxole-5-carboxaldehyde, 7-methoxy, 5-methoxypiperonal, 7-methoxy-benzo 1,3 dioxole-5-carbaldehyde, 7-methoxybenzo d 1,3 dioxole-5-carbaldehyde, 7-methoxy-2h-1,3-benzodioxole-5-carbaldehyde, 7-methoxybenzo-1,3-dioxole-5-carboxaldehyde, 7-methoxybenzo 1,3 dioxole-5-carbaldehyde
LOFRBHZYZCIOJO-UHFFFAOYSA-N
7-methoxy-1,3-benzodioxole-5-carbaldehyde
22016
97%
5780-07-4
C9H8O4
5 g
Air Sensitive
Insoluble in water.
COC1=CC(=CC2=C1OCO2)C=O
180.16
180.16

Chemical Identifiers

5780-07-4
MFCD00016900
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-304-8

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only