missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxypiperonal, 97%

Catalog No. AAL0821303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0821303 1 g
AAL0821306 5 g
AAL0821314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0821303 Supplier Thermo Scientific Chemicals Supplier No. L0821303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5780-07-4 | C9H8O4 | 180.16 g/mol

5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxypiperonal is used in the synthesis of oxazole combretastatin A-4 analogs showing anti-tumor activity. It is also used in the synthesis of endothelin-A antagonist ABT-546, and further in the synthesis of antidepressant rolipram.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with air, oxidizing agents. Store under dry inert gas. Store away from air.

Chemical Identifiers

CAS 5780-07-4
MDL Number MFCD00016900

Specifications

Melting Point 126°C to 132°C
Molecular Formula C8H10N2O2
Quantity 1 g
Beilstein 384077
Sensitivity Air Sensitive
Synonym 3-methoxybenzhydrazide, m-anisic hydrazide, m-anisohydrazide, m-methoxy benzhydrazide, 3-methoxybenzoic hydrazide, benzoylhydrazine, m-methoxy, 3-methoxybenzenecarbohydrazide, 3-methoxybenzene-1-carbohydrazide, benzoic acid, 3-methoxy-, hydrazide, m-anisoylhydrazine
Solubility Information Insoluble in water.
InChI Key VMZSDAQEWPNOIB-UHFFFAOYSA-N
SMILES COC1=CC=CC(=C1)C(=O)NN
IUPAC Name 3-methoxybenzohydrazide
Molecular Weight (g/mol) 166.18
PubChem CID 79848
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 5-Methoxypiperonal
Show More Show Less
EINECSNumber 227-304-8
TSCA Yes

RUO – Research Use Only