missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyisatin 95.0+%, TCI America™

Catalog No. p-7132688
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13625G 5 g
M136225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13625G Supplier TCI America Supplier No. M13625G
Only null left

Chemical Identifiers

CAS 39755-95-8
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00169023
InChI Key DMHGXMPXHPOXBF-UHFFFAOYSA-N
Synonym 5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci
PubChem CID 38333
IUPAC Name 5-methoxy-2,3-dihydro-1H-indole-2,3-dione
SMILES COC1=CC=C2NC(=O)C(=O)C2=C1

Specifications

Melting Point 203°C
Quantity 5 g
Formula Weight 177.16
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyisatin
RTECSNumber NL7938000
TSCA No