missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyisatin, 97%

Catalog No. p-7026641
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2274206 5 g
AAB2274214 25 g
AAB2274222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2274206 Supplier Thermo Scientific Chemicals Supplier No. B2274206
Only null left

CAS: 39755-95-8 | C9H7NO3 | 177.16 g/mol

5-Methoxyisatin is used for Pharmaceutical intermediates, a raw material used in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxyisatin is used for Pharmaceutical intermediates, a raw material used in chemical synthesis.

Solubility
Soluble in cold water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 39755-95-8
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.16
MDL Number MFCD00169023
InChI Key DMHGXMPXHPOXBF-UHFFFAOYSA-N
Synonym 5-methoxyisatin, 5-methoxyindoline-2,3-dione, 5-methoxy-2,3-indolinedione, 1h-indole-2,3-dione, 5-methoxy, 5-methoxy-indole-2,3-dione, 5-methoxy isatin, 5-methoxyindole-2,3-dione, 5-methoxy-2,3-dihydro-1h-indole-2,3-dione, indole-2,3-dione, 5-methoxy, 1h-indole-2,3-dione, 5-methoxy-9ci
PubChem CID 38333
SMILES COC1=CC=C2NC(=O)C(=O)C2=C1

Specifications

Melting Point 199°C to 202°C
Quantity 5 g
Solubility Information Soluble in cold water.
IUPAC Name 5-methoxy-1H-indole-2,3-dione
Formula Weight 177.16
Percent Purity 98%
Chemical Name or Material 5-Methoxyisatin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber NL7938000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only