missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole 99.0+%, TCI America™

Catalog No. M07311G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07311G 1 g
M07315G 5 g
M073125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M07311G Supplier TCI America Supplier No. M07311G

Chemical Identifiers

CAS 1006-94-6
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00005674
InChI Key DWAQDRSOVMLGRQ-UHFFFAOYSA-N
Synonym 5-methoxyindole, 1h-indole, 5-methoxy, femedol, 5-methoxy indole, indol-5-yl methyl ether, indole, 5-methoxy, methoxy-5 indole, unii-dqm3as43pq, methoxy-5 indole french, methoxyindole 5-
PubChem CID 13872
IUPAC Name 5-methoxy-1H-indole
SMILES COC1=CC2=C(C=C1)NC=C2

Specifications

Melting Point 55°C
Boiling Point 178°C
Quantity 1 g
Formula Weight 147.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyindole
RTECSNumber NL9500000
TSCA No
Recommended Storage Refrigerator