missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole, 99%

Catalog No. AAB2529014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2529014 25 g
AAB2529003 1 g
AAB2529006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2529014 Supplier Thermo Scientific Chemicals Supplier No. B2529014

CAS: 1006-94-6 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1006-94-6
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00005674
InChI Key DWAQDRSOVMLGRQ-UHFFFAOYSA-N
Synonym 5-methoxyindole, 1h-indole, 5-methoxy, femedol, 5-methoxy indole, indol-5-yl methyl ether, indole, 5-methoxy, methoxy-5 indole, unii-dqm3as43pq, methoxy-5 indole french, methoxyindole 5-
PubChem CID 13872
IUPAC Name 5-methoxy-1H-indole
SMILES COC1=CC2=C(C=C1)NC=C2

Specifications

Melting Point 52°C to 55°C
Boiling Point 176°C to 178°C (17 mmHg)
Quantity 25 g
Beilstein 116722
Sensitivity Light sensitive
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 5-Methoxyindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-745-3
RTECSNumber NL9500000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only