missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-2-carboxylic Acid 97.0+%, TCI America™

Catalog No. p-7132632
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12715G 5 g
M127125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12715G Supplier TCI America Supplier No. M12715G

Chemical Identifiers

CAS 4382-54-1
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00005614
InChI Key YEBJVSLNUMZXRJ-UHFFFAOYSA-N
Synonym 5-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-methoxy, 5-methoxy-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-methoxy, acide methoxy-5 indole carboxylique-2, acide methoxy-5 indole carboxylique-2 french, 5-methoxyindol-2-carboxylic acid, 5-methoxy-1h-indole-2-carboxylicacid, pubchem1702, acmc-209jvj
PubChem CID 20401
IUPAC Name 5-methoxy-1H-indole-2-carboxylic acid
SMILES COC1=CC=C2NC(=CC2=C1)C(O)=O

Specifications

Quantity 5 g
Formula Weight 191.19
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxyindole-2-carboxylic Acid
RTECSNumber NL6005000
TSCA No