missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxyindole-2-carboxylic acid, 97%

Catalog No. AAL1513706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1513706 5 g
AAL1513703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1513706 Supplier Thermo Scientific Chemicals Supplier No. L1513706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4382-54-1 | C10H9NO3 | 191.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4382-54-1
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00005614
InChI Key YEBJVSLNUMZXRJ-UHFFFAOYSA-N
Synonym 5-methoxyindole-2-carboxylic acid, 1h-indole-2-carboxylic acid, 5-methoxy, 5-methoxy-2-indolecarboxylic acid, indole-2-carboxylic acid, 5-methoxy, acide methoxy-5 indole carboxylique-2, acide methoxy-5 indole carboxylique-2 french, 5-methoxyindol-2-carboxylic acid, 5-methoxy-1h-indole-2-carboxylicacid, pubchem1702, acmc-209jvj
PubChem CID 20401
IUPAC Name 5-methoxy-1H-indole-2-carboxylic acid
SMILES COC1=CC=C2NC(=CC2=C1)C(O)=O

Specifications

Melting Point ∽200°C (decomposition)
Quantity 5 g
Beilstein 157478
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material 5-Methoxyindole-2-carboxylic acid
EINECSNumber 224-487-6
RTECSNumber NL6005000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only