missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-7-methylindole, 97%

Catalog No. AAH6312306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6312306 5 g
AAH6312303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6312306 Supplier Thermo Scientific Chemicals Supplier No. H6312306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 61019-05-4 | C10H11NO | 161.204 g/mol

5-Methoxy-7-methylindole is employed as a intermediate for chemical research and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methoxy-7-methylindole is employed as a intermediate for chemical research and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 61019-05-4
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD09027022
InChI Key YGPVRHHGKCQSIL-UHFFFAOYSA-N
Synonym 5-methoxy-7-methylindole, 1h-indole, 5-methoxy-7-methyl, ksc612k7l, 5-methoxy7-methyl-1h-indole
PubChem CID 12338917
IUPAC Name 5-methoxy-7-methyl-1H-indole
SMILES CC1=C2C(=CC(=C1)OC)C=CN2

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 161.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Methoxy-7-methylindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only