missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-methylbenzoselenazole, 97%

Catalog No. AAB2241103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2241103 1 g
AAB2241106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2241103 Supplier Thermo Scientific Chemicals Supplier No. B2241103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2946-17-0 | C9H9NOSe | 226.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2946-17-0
Molecular Formula C9H9NOSe
Molecular Weight (g/mol) 226.148
MDL Number MFCD00005558
InChI Key LVIYTMFDCXRSDL-UHFFFAOYSA-N
Synonym 5-methoxy-2-methylbenzoselenazole, benzoselenazole, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzselenazole, 2-methyl-5-methoxybenzoselenazole, 5-methoxy-2-methylbenzo d 1,3-selenazole, #, 5-methoxy-2-methylbenzo d 1,3 selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazoline
PubChem CID 76274
IUPAC Name 5-methoxy-2-methyl-1,3-benzoselenazole
SMILES CC1=NC2=C([Se]1)C=CC(=C2)OC

Specifications

Density 1.49
Boiling Point 115°C to 117°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.639
Quantity 1 g
UN Number UN3440
Formula Weight 226.14
Percent Purity 97%
Chemical Name or Material 5-Methoxy-2-methylbenzoselenazole
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 220-956-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only