missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-2-benzimidazolinone, 98%

Catalog No. AAB2258003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2258003 1 g
AAB2258006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2258003 Supplier Thermo Scientific Chemicals Supplier No. B2258003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2080-75-3 | C8H8N2O2 | 164.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2080-75-3
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD00051946
InChI Key AUPLVAKFTYFHTA-UHFFFAOYSA-N
Synonym 5-methoxy-2-benzimidazolinone, 5-methoxy-1h-benzo d imidazol-2 3h-one, 2-hydroxy-5-methoxybenzimidazole, 5-methoxy-2-hydroxybenzimidazole, 2h-benzimidazol-2-one, 1,3-dihydro-5-methoxy, 5-methoxy-benzimidazolin-2-one, 5-methoxy-1,3-dihydro-benzoimidazol-2-one, 5-methoxy-1h-1,3-benzodiazol-2-ol, 1,3-dihydro-5-methoxy-2h-benzimidazol-2-one, 1,3-dihydro-5-methoxy-2h-benzimidazole-2-one
PubChem CID 591101
IUPAC Name 5-methoxy-1,3-dihydrobenzimidazol-2-one
SMILES COC1=CC2=C(C=C1)NC(=O)N2

Specifications

Melting Point ∼255°C (decomposition)
Quantity 1 g
Beilstein 141043
Formula Weight 164.17
Percent Purity 98%
Chemical Name or Material 5-Methoxy-2-benzimidazolinone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only