missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-1-indanone 98.0+%, TCI America™

Catalog No. p-7132753
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
M14675G 5 g
M146725G 25 g
2 options

Catalog No. M14675G

Supplier: TCI America M14675G

Only null left

Chemical Identifiers

CAS 5111-70-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00003789
InChI Key QOPRWBRNMPANKN-UHFFFAOYSA-N
Synonym 5-methoxy-1-indanone, 5-methoxyindan-1-one, 5-methoxy-2,3-dihydro-1h-inden-1-one, 5-methoxyindanone, 2,3-dihydro-5-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-5-methoxy, 5-methoxy indanone, 5-methoxyhydrindone, pubchem2031, 1-oxo-5-methoxyindane
PubChem CID 78787
IUPAC Name 5-methoxy-2,3-dihydroinden-1-one
SMILES COC1=CC2=C(C=C1)C(=O)CC2

Specifications

Melting Point 112°C
Boiling Point 150°C
Quantity 5 g
Formula Weight 162.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methoxy-1-indanone