missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-1-indanone, 98%

Catalog No. p-7045565
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1116503 1 g
AAL1116506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1116503 Supplier Thermo Scientific Chemicals Supplier No. L1116503
Only null left

CAS: 5111-70-6 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5111-70-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00003789
InChI Key QOPRWBRNMPANKN-UHFFFAOYSA-N
Synonym 5-methoxy-1-indanone, 5-methoxyindan-1-one, 5-methoxy-2,3-dihydro-1h-inden-1-one, 5-methoxyindanone, 2,3-dihydro-5-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-5-methoxy, 5-methoxy indanone, 5-methoxyhydrindone, pubchem2031, 1-oxo-5-methoxyindane
PubChem CID 78787
IUPAC Name 5-methoxy-2,3-dihydroinden-1-one
SMILES COC1=CC2=C(C=C1)C(=O)CC2

Specifications

Melting Point 108°C to 110°C
Quantity 1 g
Beilstein 1282137
Formula Weight 162.19
Percent Purity 98%
Chemical Name or Material 5-Methoxy-1-indanone
Hazard Category H302+H312+H332
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 225-838-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only