missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic Acid 97.0+%, TCI America™
$126.76 - $642.54
Chemical Identifiers
| CAS | 53723-88-9 |
|---|---|
| Molecular Formula | C4H4N2O2S3 |
| Molecular Weight (g/mol) | 208.268 |
| MDL Number | MFCD00129970 |
| InChI Key | UJBXVTJYSIDCIE-UHFFFAOYSA-N |
| Synonym | 5-mercapto-1,3,4-thiadiazol-2-ylthio acetic acid, 2-5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid, 5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid, 4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio acetic acid, acetic acid, 4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio, 5-mercapto-1,3,4-thiadiazole-2-ylthio acetic acid, 5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid, 5-mercapto-1,3,4-thiadiazole-2-ylthio aceticacid, 2-5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid, 2-5-sulfanyl-1,3,4-thiadiazol-2-ylthio acetic acid |
| PubChem CID | 1714399 |
| IUPAC Name | 2-[(2-sulfanylidene-3H-1,3,4-thiadiazol-5-yl)sulfanyl]acetic acid |
| SMILES | C(C(=O)O)SC1=NNC(=S)S1 |
Chemical Identifiers
| 53723-88-9 | |
| 208.268 | |
| UJBXVTJYSIDCIE-UHFFFAOYSA-N | |
| 1714399 | |
| C(C(=O)O)SC1=NNC(=S)S1 |
| C4H4N2O2S3 | |
| MFCD00129970 | |
| 5-mercapto-1,3,4-thiadiazol-2-ylthio acetic acid, 2-5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid, 5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid, 4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio acetic acid, acetic acid, 4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio, 5-mercapto-1,3,4-thiadiazole-2-ylthio acetic acid, 5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid, 5-mercapto-1,3,4-thiadiazole-2-ylthio aceticacid, 2-5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid, 2-5-sulfanyl-1,3,4-thiadiazol-2-ylthio acetic acid | |
| 2-[(2-sulfanylidene-3H-1,3,4-thiadiazol-5-yl)sulfanyl]acetic acid |
Specifications
| 53723-88-9 | |
| Yellow | |
| MFCD00129970 | |
| 5-mercapto-1,3,4-thiadiazol-2-ylthio acetic acid, 2-5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid, 5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid, 4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio acetic acid, acetic acid, 4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio, 5-mercapto-1,3,4-thiadiazole-2-ylthio acetic acid, 5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid, 5-mercapto-1,3,4-thiadiazole-2-ylthio aceticacid, 2-5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid, 2-5-sulfanyl-1,3,4-thiadiazol-2-ylthio acetic acid | |
| C(C(=O)O)SC1=NNC(=S)S1 | |
| 208.268 | |
| 208.27 | |
| Crystalline Powder |
| 167°C | |
| C4H4N2O2S3 | |
| 5 g | |
| UJBXVTJYSIDCIE-UHFFFAOYSA-N | |
| 2-[(2-sulfanylidene-3H-1,3,4-thiadiazol-5-yl)sulfanyl]acetic acid | |
| 1714399 | |
| ≥97.0% (HPLC,T) | |
| (5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic Acid |
Safety and Handling
TSCA : No