missing translation for 'onlineSavingsMsg'
Learn More

5-Ketohexanenitrile 98.0+%, TCI America™

Catalog No. k002925ml
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K002925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K002925ML Supplier TCI America Supplier No. K002925ML
Only null left

Chemical Identifiers

CAS 10412-98-3
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.144
MDL Number MFCD00010671
InChI Key AEVMBQIIZGKQRB-UHFFFAOYSA-N
Synonym 4-Acetylbutyronitrile, 5-Oxohexanenitrile
PubChem CID 82611
IUPAC Name 5-oxohexanenitrile
SMILES CC(=O)CCCC#N

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 mL
Formula Weight 111.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Ketohexanenitrile
EINECSNumber (2)-1547
TSCA Yes