missing translation for 'onlineSavingsMsg'
Learn More

5-Keto-D-gluconic acid potassium salt, 98%

Catalog No. AAJ6691606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6691606 5 g
AAJ6691603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6691606 Supplier Thermo Scientific Chemicals Supplier No. J6691606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5447-60-9 | C6H10KO7 | 233.237 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Soluble in water 50mg/ml

Notes
Store at -20°C. Incompatible with heat, oxidizing agents.

Chemical Identifiers

Molecular Formula C6H10KO7
Molecular Weight (g/mol) 233.237
MDL Number MFCD00069562
InChI Key BJBNFUCYWPVFKM-YMDUGQBDSA-N
Synonym potassium 5-ketogluconate
PubChem CID 131855000
IUPAC Name potassium;(2R,3S,4S)-2,3,4,6-tetrahydroxy-5-oxohexanoic acid
SMILES C(C(=O)C(C(C(C(=O)O)O)O)O)O.[K]

Specifications

Color White
Quantity 5 g
Solubility Information Soluble in water 50mg/ml
Formula Weight 232.23
Percent Purity 0.98
Physical Form Powder
Chemical Name or Material 5-Keto-D-gluconic acid potassium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only