missing translation for 'onlineSavingsMsg'
Learn More

Lemon Oil, California Type, Cold-pressed, FCC, 2.2-3.8%, Spectrum™ Chemical

Catalog No. p-7157672
Change view
Click to view available options
Quantity:
500 mL
15 kg
Packaging:
Amber Glass Bottle
Steel Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603775 500 mL Amber Glass Bottle
18603776 15 kg Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18603775 Supplier Spectrum Chemical Mfg Cor Supplier No. L1200500MLGL

L1200, 8008-56-8

Spectrum™ Chemical Lemon Oil, California Type, Cold-pressed, FCC is used in aromatherapy and as a flavorant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8008-56-8

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Refractive Index 1.473-1.476
Quantity 500 mL
UN Number UN1197
Percent Purity 2.2 to 3.8%
Grade FCC