missing translation for 'onlineSavingsMsg'
Learn More

5-Iodovanillin, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 156670050

 View more versions of this product

Catalog No. AC156670050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

5-Iodovanillin
5438-36-8
100.0
96% min. (HPLC)
C8H7IO3
MFCD00006941
08, 260
FBBCSYADXYILEH-UHFFFAOYSA-N
4-hydroxy-3-iodo-5-methoxybenzaldehyde
79499
97%
97%
96.0
Authentic
Glass bottle
IC6H2(OCH3)(OH)CHO
5g
5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
COC1=C(C(=CC(=C1)C=O)I)O
278.04
278.04

Chemical Identifiers

5438-36-8
278.04
FBBCSYADXYILEH-UHFFFAOYSA-N
79499
COC1=C(C(=CC(=C1)C=O)I)O
C8H7IO3
MFCD00006941
5-iodovanillin, 5 iodovanillin, benzaldehyde, 4-hydroxy-3-iodo-5-methoxy, 5-iodo vanillin, 3-methoxy-4-hydroxy-5-iodobenzaldehyde, 5-iodovanilline, 5-iodovanilin, 5-iodo vaniline, acmc-1ao68
4-hydroxy-3-iodo-5-methoxybenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-617-7

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only