missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 5-Iodouracil, 99%

CAS: 696-07-1 | C4H3IN2O2 | 237.98 g/mol

$216.72 - $216.72

Chemical Identifiers

CAS 696-07-1
Molecular Formula C4H3IN2O2
Molecular Weight (g/mol) 237.98
MDL Number MFCD00006020
InChI Key KSNXJLQDQOIRIP-UHFFFAOYSA-N
Synonym 5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 69672
ChEBI CHEBI:43636
IUPAC Name 5-iodo-1H-pyrimidine-2,4-dione
SMILES IC1=CNC(=O)NC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC228160100
SDS View Documents
Thermo Scientific Chemicals
228160100
10 g Glass bottle
Each for $216.72
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

696-07-1
237.98
KSNXJLQDQOIRIP-UHFFFAOYSA-N
69672
5-iodo-1H-pyrimidine-2,4-dione
C4H3IN2O2
MFCD00006020
5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
CHEBI:43636
IC1=CNC(=O)NC1=O

Specifications

696-07-1
100.0
White to Yellow
99%
C4H3IN2O2
10 g
KSNXJLQDQOIRIP-UHFFFAOYSA-N
5-iodo-1H-pyrimidine-2,4-dione
69672
237.98
Fluffy Powder
98.5
270.0°C to 274.0°C
Authentic
Glass bottle
MFCD00006020
5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione
IC1=CNC(=O)NC1=O
237.98
CHEBI:43636
99%
5-Iodouracil
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only