Learn More
5-Iodouracil, 97%
CAS: 696-07-1 | C4H3IN2O2 | 237.98 g/mol
Supplier: Thermo Scientific Chemicals A1899409
Description
It is used as an intermediate. It has an antitumor activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as an intermediate. It has an antitumor activity.
Solubility
Very faint turbidity in NH3aq. Soluble in 1M NaOH.
Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| 5-Iodouracil | |
| 274°C to 276°C | |
| MFCD00006020 | |
| 4891 | |
| 5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| KSNXJLQDQOIRIP-UHFFFAOYSA-N | |
| 5-iodo-1H-pyrimidine-2,4-dione | |
| 69672 | |
| 237.99 |
| 696-07-1 | |
| C4H3IN2O2 | |
| 10 g | |
| Light sensitive | |
| Very faint turbidity in NH3aq. Soluble in 1M NaOH. | |
| IC1=CNC(=O)NC1=O | |
| 237.98 | |
| CHEBI:43636 | |
| 97% |
Chemical Identifiers
| 696-07-1 | |
| 237.98 | |
| KSNXJLQDQOIRIP-UHFFFAOYSA-N | |
| 69672 | |
| 5-iodo-1H-pyrimidine-2,4-dione |
| C4H3IN2O2 | |
| MFCD00006020 | |
| 5-iodouracil, 5-iodopyrimidine-2,4 1h,3h-dione, 2,4 1h,3h-pyrimidinedione, 5-iodo, uracil, 5-iodo, 5-iodopyrimidine-2,4-diol, 2,4-dihydroxy-5-iodopyrimidine, 5-iodo uracil, unii-h59brk500m, chembl1173, 5-iodo-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| CHEBI:43636 | |
| IC1=CNC(=O)NC1=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 211-788-2
RTECSNumber : YR0525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only