missing translation for 'onlineSavingsMsg'
Learn More

5-Iodoisatin 98.0+%, TCI America™

Supplier:  TCI America I06405G

Catalog No. I06405G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Iodoisatin
C8H4INO2
5-iodoisatin, 5-iodoindoline-2,3-dione, 5-iodo isatin, 5-iodoindole-2,3-dione, 5-iodo-2,3-dioxoindoline, 1h-indole-2,3-dione, 5-iodo, 5-iodo-2,3-dihydro-1h-indole-2,3-dione, 5-iodo-1h-benzo d azolidine-2,3-dione, 5-iodo-isatine, 5-iodo-isatin
C1=CC2=C(C=C1I)C(=O)C(=O)N2
273.029
273.03
Crystalline Powder
20780-76-1
5 g
OEUGDMOJQQLVAZ-UHFFFAOYSA-N
5-iodo-1H-indole-2,3-dione
88695
≥98.0% (T)

Chemical Identifiers

20780-76-1
273.029
5-iodoisatin, 5-iodoindoline-2,3-dione, 5-iodo isatin, 5-iodoindole-2,3-dione, 5-iodo-2,3-dioxoindoline, 1h-indole-2,3-dione, 5-iodo, 5-iodo-2,3-dihydro-1h-indole-2,3-dione, 5-iodo-1h-benzo d azolidine-2,3-dione, 5-iodo-isatine, 5-iodo-isatin
5-iodo-1H-indole-2,3-dione
C8H4INO2
OEUGDMOJQQLVAZ-UHFFFAOYSA-N
88695
C1=CC2=C(C=C1I)C(=O)C(=O)N2
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions