missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Iodoindole, 98%
Description
5-Iodoindole is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is used as the starting material in the synthesis of 5-(α-fluorovinyl)-N-tosylindole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Iodoindole is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is used as the starting material in the synthesis of 5-(α-fluorovinyl)-N-tosylindole.
Notes
Store at room temperature. Incompatible with strong oxidizing agents. Light sensitive.
5-Iodoindole is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is used as the starting material in the synthesis of 5-(α-fluorovinyl)-N-tosylindole.
Notes
Store at room temperature. Incompatible with strong oxidizing agents. Light sensitive.
Specifications
Specifications
| Melting Point | 97°C to 100°C |
| Quantity | 5 g |
| Sensitivity | Light sensitive |
| Formula Weight | 243.05 |
| Percent Purity | 98% |
| Chemical Name or Material | 5-Iodoindole |
Safety and Handling
| Hazard Category | H302+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only