missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-phenoxypyridine, ≥95%, Thermo Scientific™

Catalog No. p-2702768
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC19611DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC19611DA Supplier Thermo Scientific Chemicals Supplier No. CC19611DA

Chemical Identifiers

CAS 352018-92-9
Molecular Formula C11H8INO
Molecular Weight (g/mol) 297.095
MDL Number MFCD02681952
InChI Key JCEXQQNAXWYXRX-UHFFFAOYSA-N
Synonym 5-iodo-2-phenoxy-pyridine, pyridine,5-iodo-2-phenoxy
PubChem CID 2776503
IUPAC Name 5-iodo-2-phenoxypyridine
SMILES C1=CC=C(C=C1)OC2=NC=C(C=C2)I

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 297.09
Percent Purity 95%
Chemical Name or Material 5-Iodo-2-phenoxypyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.