missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-methoxypyridine 98.0+%, TCI America™

Catalog No. I09251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09251G 1 g
I09255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09251G Supplier TCI America Supplier No. I09251G
Only null left

Chemical Identifiers

CAS 13472-61-2
Molecular Formula C6H6INO
Molecular Weight (g/mol) 235.024
MDL Number MFCD07781180
InChI Key NTXRNCUPGYOZCN-UHFFFAOYSA-N
PubChem CID 23423786
IUPAC Name 5-iodo-2-methoxypyridine
SMILES COC1=NC=C(C=C1)I

Specifications

Boiling Point 74°C
Quantity 1 g
Synonym 2-methoxy-5-iodopyridine, pyridine, 5-iodo-2-methoxy, pyridine,5-iodo-2-methoxy, pubchem6606, acmc-1c1li, 5-iodanyl-2-methoxy-pyridine
Formula Weight 235.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Iodo-2-methoxypyridine