missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Iodo-2-methoxypyridine, 97%, Thermo Scientific™
Specifications
Specifications
| CAS Min % | 96.0 |
| CAS Max % | 100.0 |
| Boiling Point | 106°C to 108°C (30.0 Torr) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 1g |
| Formula Weight | 235.03 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Iodo-2-methoxypyridine |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
RUO – Research Use Only