missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-furaldehyde 98.0+%, TCI America™

Catalog No. I06961G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06961G 1 g
I06965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06961G Supplier TCI America Supplier No. I06961G

Chemical Identifiers

CAS 2689-65-8
Molecular Formula C5H3IO2
Molecular Weight (g/mol) 221.981
MDL Number MFCD00159503
InChI Key QPGPCPKDVSPJAY-UHFFFAOYSA-N
Synonym 5-iodo-2-furaldehyde, 5-iodofurfural, 5-iodo-2-furancarboxaldehyde, 5-iodo-2-furaldehdye, 2-furancarboxaldehyde, 5-iodo, 5-iodofuraldehyde, acmc-209gtv, 5-iodofurancarboxaldehyde, 5-iodanylfuran-2-carbaldehyde
PubChem CID 693264
IUPAC Name 5-iodofuran-2-carbaldehyde
SMILES C1=C(OC(=C1)I)C=O

Specifications

Melting Point 128°C
Color Yellow
Quantity 1 g
Formula Weight 221.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Iodo-2-furaldehyde